4-Fluoro-2-methoxyphenylacetonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Fluoro-2-methoxyphenyl)acetonitrile

PC408265-1G
APOSPC408265-1GEA 55.1 EUR
APOSPC408265-1G
4-Fluoro-2-methoxyphenylacetonitrile
4-Fluoro-2-methoxyphenylacetonitrile
Formel: C₉H₈FNO
Molecular Weight: 165.164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660192
CAS-Nummer: 886498-56-2
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR