3-(Trifluoromethoxy)phenoxyacetic acid

Lieferant: Apollo Scientific
Achtung

PC408280-1G
APOSPC408280-1GEA 176 EUR
APOSPC408280-1G
3-(Trifluoromethoxy)phenoxyacetic acid
3-(Trifluoromethoxy)phenoxyacetic acid
Formel: C₉H₇F₃O₄
Molecular Weight: 236,145 g/mol
Schmelzpunkt: 60…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD19105099
CAS-Nummer: 836-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR