3-Amino-5-fluorbenzoesäure 97%
Lieferant: Apollo Scientific
PC408286-25G
APOSPC408286-25GEA
224
EUR
APOSPC408286-25G
3-Amino-5-fluorbenzoesäure 97%
3-Amino-5-fluorobenzoic acid
Formel:
C₇H₆FNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07779526 CAS-Nummer: 786616-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...