2-Fluoro-5-iodo-4-methylpyridine 97%

Lieferant: Apollo Scientific

PC408288-1G
APOSPC408288-1GEA 156 EUR
APOSPC408288-1G
2-Fluoro-5-iodo-4-methylpyridine 97%
2-Fluoro-5-iodo-4-methylpyridine
Formel: C₆H₅FIN
Molecular Weight: 237,013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12911585
CAS-Nummer: 1184913-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR