5-Fluoro-N-isopropylpyridin-2-amine

Lieferant: Apollo Scientific

PC408292-1G
APOSPC408292-1GEA 1200 EUR
APOSPC408292-1G
5-Fluoro-N-isopropylpyridin-2-amine
5-Fluoro-N-isopropylpyridin-2-amine
Formel: C₈H₁₁FN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14621285
CAS-Nummer: 1251039-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR