(5-Fluoro-pyrimidin-2-yl)-isopropyl-amine

Lieferant: Apollo Scientific

PC408293-1G
APOSPC408293-1GEA 430 EUR
APOSPC408293-1G
(5-Fluoro-pyrimidin-2-yl)-isopropyl-amine
(5-Fluoro-pyrimidin-2-yl)-isopropyl-amine
Formel: C₇H₁₀FN₃
Molecular Weight: 155,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02102214
CAS-Nummer: 77476-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR