4-(4-Fluoro-benzenesulphonylmethyl)-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408294-500MG
APOSPC408294-500MGEA 495 EUR
APOSPC408294-500MG
4-(4-Fluoro-benzenesulphonylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
4-(4-Fluoro-benzenesulphonylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₄FNO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD21098431
CAS-Nummer: 333988-27-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR