2-(4-Fluoro-phenylsulphanylmethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408295-500MG
APOSPC408295-500MGEA 430 EUR
APOSPC408295-500MG
2-(4-Fluoro-phenylsulphanylmethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
2-(4-Fluoro-phenylsulphanylmethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Formel: C₁₆H₂₂FNO₂S
Molecular Weight: 311,415 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098435
CAS-Nummer: 1353945-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR