(S)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine hydrochloride

Lieferant: Apollo Scientific

PC408296-500MG
APOSPC408296-500MGEA 495 EUR
APOSPC408296-500MG
(S)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine hydrochloride
(S)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine hydrochloride
Formel: C₁₀H₁₂FNO₂S·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD21098440
CAS-Nummer: 1354010-06-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR