3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ylamine hydrochloride
Lieferant: Apollo Scientific
PC408299-500MG
APOSPC408299-500MGEA
553
EUR
APOSPC408299-500MG
3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ylamine hydrochloride
3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ylamine hydrochloride
Formel:
C₁₁H₁₃ClF₃N₃·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD09877740 CAS-Nummer: 1185313-62-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...