Methyl-5-bromo-3-(trifluoromethyl)picolinate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-bromo-3-(trifluoromethyl)picolinate, Methyl 5-bromo-3-(trifluoromethyl)-2-pyridinecarboxylate
PC408300-250MG
APOSPC408300-250MGEA
108
EUR
APOSPC408300-250MG
Methyl-5-bromo-3-(trifluoromethyl)picolinate 98%
Methyl-5-bromo-3-(trifluoromethyl)picolinate
Formel:
C₈H₅BrF₃NO₂ Molecular Weight: 284,0300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14698001 CAS-Nummer: 1214328-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...