(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-piperidin-4-yl-amine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-N-(piperidin-4-yl)-5-(trifluoromethyl)pyridin-2-amine hydrochloride

PC408302-500MG
APOSPC408302-500MGEA 553 EUR
APOSPC408302-500MG
(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-piperidin-4-yl-amine hydrochloride
(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-piperidin-4-yl-amine hydrochloride
Formel: C₁₁H₁₃ClF₃N₃·ClH
Molecular Weight: 316,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698911
CAS-Nummer: 1185315-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR