1-(2-Fluoro-benzyl)-piperidin-3-ol
Lieferant: Apollo Scientific
Synonyme:
1-(2-Fluorobenzyl)piperidin-3-ol
PC408303-500MG
APOSPC408303-500MGEA
430
EUR
APOSPC408303-500MG
1-(2-Fluoro-benzyl)-piperidin-3-ol
1-(2-Fluoro-benzyl)-piperidin-3-ol
Formel:
C₁₂H₁₆FNO Molecular Weight: 209,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01306500 CAS-Nummer: 864388-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...