[1-(2-Fluoro-benzyl)-piperidin-4-yl]-methanol

Lieferant: Apollo Scientific

Synonyme: (1-(2-Fluorobenzyl)piperidin-4-yl)methanol

PC408304-500MG
APOSPC408304-500MGEA 430 EUR
APOSPC408304-500MG
[1-(2-Fluoro-benzyl)-piperidin-4-yl]-methanol
[1-(2-Fluoro-benzyl)-piperidin-4-yl]-methanol
Formel: C₁₃H₁₈FNO
Molecular Weight: 223,287 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16153520
CAS-Nummer: 1241253-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR