4-[(3-Fluorobenzyl)oxy]piperidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-((3-Fluorobenzyl)oxy)piperidine hydrochloride

PC408305-500MG
APOSPC408305-500MGEA 430 EUR
APOSPC408305-500MG
4-[(3-Fluorobenzyl)oxy]piperidine hydrochloride
4-[(3-Fluorobenzyl)oxy]piperidine hydrochloride
Formel: C₁₂H₁₇ClFNO
Molecular Weight: 245,73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11506394
CAS-Nummer: 1185298-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR