1-(4-Fluoro-benzyl)-piperidine-4-carboxylic acid 4-fluoro-benzyl ester
Lieferant: Apollo Scientific
Synonyme:
4-Fluorobenzyl 1-(4-Fluorobenzyl)piperidine-4-carboxylate
PC408306-500MG
APOSPC408306-500MGEA
430
EUR
APOSPC408306-500MG
1-(4-Fluoro-benzyl)-piperidine-4-carboxylic acid 4-fluoro-benzyl ester
1-(4-Fluoro-benzyl)-piperidine-4-carboxylic acid 4-fluoro-benzyl ester
Formel:
C₂₀H₂₁F₂NO₂ Molecular Weight: 345,383 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18379928 CAS-Nummer: 1261230-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...