4-[(2-Fluorobenzyl)oxy]piperidine hydrochloride
Lieferant: Apollo Scientific
PC408307-500MG
APOSPC408307-500MGEA
430
EUR
APOSPC408307-500MG
4-[(2-Fluorobenzyl)oxy]piperidine hydrochloride
4-[(2-Fluorobenzyl)oxy]piperidine hydrochloride
Formel:
C₁₂H₁₇ClFNO Molecular Weight: 245,73 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13561002 CAS-Nummer: 614731-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...