(2-Fluoro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride

Lieferant: Apollo Scientific

PC408309-500MG
APOSPC408309-500MGEA 430 EUR
APOSPC408309-500MG
(2-Fluoro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride
(2-Fluoro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride
Formel: C₁₃H₁₉FN₂·ClH
Molecular Weight: 258,763 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18380005
CAS-Nummer: 1261235-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR