(3-Fluoro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride

Lieferant: Apollo Scientific

Synonyme: N-(3-Fluorobenzyl)-N-methylpiperidin-4-amine hydrochloride

PC408310-500MG
APOSPC408310-500MGEA 430 EUR
APOSPC408310-500MG
(3-Fluoro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride
(3-Fluoro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride
Formel: C₁₃H₁₉FN₂·ClH
Molecular Weight: 258,763 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18380007
CAS-Nummer: 1261231-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR