(4-Fluoro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride

Lieferant: Apollo Scientific

PC408311-500MG
APOSPC408311-500MGEA 430 EUR
APOSPC408311-500MG
(4-Fluoro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride
(4-Fluoro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride
Formel: C₁₃H₁₉FN₂·ClH
Molecular Weight: 258,763 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18380009
CAS-Nummer: 1261230-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR