[1-(5-Fluoro-pyrimidin-2-yl)-piperidin-3-yl]-methanol

Lieferant: Apollo Scientific

Synonyme: (1-(5-Fluoropyrimidin-2-yl)piperidin-3-yl)methanol

PC408312-500MG
APOSPC408312-500MGEA 553 EUR
APOSPC408312-500MG
[1-(5-Fluoro-pyrimidin-2-yl)-piperidin-3-yl]-methanol
[1-(5-Fluoro-pyrimidin-2-yl)-piperidin-3-yl]-methanol
Formel: C₁₀H₁₄FN₃O
Molecular Weight: 211,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18380078
CAS-Nummer: 1261232-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR