3-(5-Fluoro-pyrimidin-2-yloxy)-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408313-500MG
APOSPC408313-500MGEA 553 EUR
APOSPC408313-500MG
3-(5-Fluoro-pyrimidin-2-yloxy)-piperidine-1-carboxylic acid tert-butyl ester
3-(5-Fluoro-pyrimidin-2-yloxy)-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₄H₂₀FN₃O₃
Molecular Weight: 297,325 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18380085
CAS-Nummer: 1261230-08-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR