(1-(5-Fluoropyrimidin-2-yl)piperidin-4-yl)methanol

Lieferant: Apollo Scientific

PC408314-500MG
APOSPC408314-500MGEA 553 EUR
APOSPC408314-500MG
(1-(5-Fluoropyrimidin-2-yl)piperidin-4-yl)methanol
(1-(5-Fluoropyrimidin-2-yl)piperidin-4-yl)methanol
Formel: C₁₀H₁₄FN₃O
Molecular Weight: 211,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18380094
CAS-Nummer: 1032825-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR