5-Fluoro-2-(pyrrolidin-2-ylmethoxy)-pyrimidine hydrochloride

Lieferant: Apollo Scientific

PC408315-500MG
APOSPC408315-500MGEA 553 EUR
APOSPC408315-500MG
5-Fluoro-2-(pyrrolidin-2-ylmethoxy)-pyrimidine hydrochloride
5-Fluoro-2-(pyrrolidin-2-ylmethoxy)-pyrimidine hydrochloride
Formel: C₉H₁₂FN₃O·ClH
Molecular Weight: 233,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18380140
CAS-Nummer: 1261235-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR