1-(4-Fluoro-benzyl)-piperidine-4-carboxylic acid
Lieferant: Apollo Scientific
PC408316-500MG
APOSPC408316-500MGEA
429
EUR
APOSPC408316-500MG
1-(4-Fluoro-benzyl)-piperidine-4-carboxylic acid
1-(4-Fluoro-benzyl)-piperidine-4-carboxylic acid
Formel:
C₁₃H₁₆FNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03371459 CAS-Nummer: 193538-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...