1-(4-Fluorobenzyl)piperidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC408317-500MG
APOSPC408317-500MGEA 306 EUR
APOSPC408317-500MG
1-(4-Fluorobenzyl)piperidine-3-carboxylic acid
1-(4-Fluorobenzyl)piperidine-3-carboxylic acid
Formel: C₁₃H₁₆FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04971127
CAS-Nummer: 832737-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR