1-(4-Fluorobenzyl)piperidine-2-carboxylic acid
Lieferant: Apollo Scientific
PC408318-500MG
APOSPC408318-500MGEA
306
EUR
APOSPC408318-500MG
1-(4-Fluorobenzyl)piperidine-2-carboxylic acid
1-(4-Fluorobenzyl)piperidine-2-carboxylic acid
Formel:
C₁₃H₁₆FNO₂ Molecular Weight: 237,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05174530 CAS-Nummer: 1030610-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...