1-(5-Fluoro-pyrimidin-2-yl)-azetidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC408319-500MG
APOSPC408319-500MGEA 619 EUR
APOSPC408319-500MG
1-(5-Fluoro-pyrimidin-2-yl)-azetidine-3-carboxylic acid
1-(5-Fluoro-pyrimidin-2-yl)-azetidine-3-carboxylic acid
Formel: C₈H₈FN₃O₂
Molecular Weight: 197,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837436
CAS-Nummer: 1289387-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR