1-(2-Chloro-6-fluoro-benzyl)-azetidine-3-carboxylic acid 2-chloro-6-fluoro-benzyl ester

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-6-fluorobenzyl 1-(2-chloro-6-fluorobenzyl)azetidine-3-carboxylate

PC408320-500MG
APOSPC408320-500MGEA 430 EUR
APOSPC408320-500MG
1-(2-Chloro-6-fluoro-benzyl)-azetidine-3-carboxylic acid 2-chloro-6-fluoro-benzyl ester
1-(2-Chloro-6-fluoro-benzyl)-azetidine-3-carboxylic acid 2-chloro-6-fluoro-benzyl ester
Formel: C₁₈H₁₅Cl₂F₂NO₂
Molecular Weight: 386,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837439
CAS-Nummer: 1289385-42-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR