1-(3-Fluoro-benzyl)-piperazine-2-carboxylic acid hydrochloride
Lieferant: Apollo Scientific
PC408322-500MG
APOSPC408322-500MGEA
430
EUR
APOSPC408322-500MG
1-(3-Fluoro-benzyl)-piperazine-2-carboxylic acid hydrochloride
1-(3-Fluoro-benzyl)-piperazine-2-carboxylic acid hydrochloride
Formel:
C₁₂H₁₅FN₂O₂·ClH Molecular Weight: 274,719 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18837468 CAS-Nummer: 1289384-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...