3-(5-Fluoro-pyrimidin-2-ylamino)-pyrrolidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 3-((5-fluoropyrimidin-2-yl)amino)pyrrolidine-1-carboxylate
PC408323-500MG
APOSPC408323-500MGEA
619
EUR
APOSPC408323-500MG
3-(5-Fluoro-pyrimidin-2-ylamino)-pyrrolidine-1-carboxylic acid tert-butyl ester
3-(5-Fluoro-pyrimidin-2-ylamino)-pyrrolidine-1-carboxylic acid tert-butyl ester
Formel:
C₁₃H₁₉FN₄O₂ Molecular Weight: 282,314 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18837486 CAS-Nummer: 1289385-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...