4-(4-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-(((4-Fluorophenyl)thio)methyl)piperidine hydrochloride

PC408324-500MG
APOSPC408324-500MGEA 430 EUR
APOSPC408324-500MG
4-(4-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
4-(4-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
Formel: C₁₂H₁₆FNS·ClH
Molecular Weight: 261,786 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837521
CAS-Nummer: 1289384-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR