4-(4-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-(((4-fluorophenyl)thio)methyl)piperidine-1-carboxylate

PC408325-500MG
APOSPC408325-500MGEA 430 EUR
APOSPC408325-500MG
4-(4-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
4-(4-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₄FNO₂S
Molecular Weight: 325.441 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837522
CAS-Nummer: 333988-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR