4-(4-Fluoro-phenylsulphanyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC408327-500MG
APOSPC408327-500MGEA 430 EUR
APOSPC408327-500MG
4-(4-Fluoro-phenylsulphanyl)-piperidine hydrochloride
4-(4-Fluoro-phenylsulphanyl)-piperidine hydrochloride
Formel: C₁₁H₁₄FNS·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD04117760
CAS-Nummer: 101798-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR