4-(4-Fluoro-benzenesulphonyl)-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408328-500MG
APOSPC408328-500MGEA 430 EUR
APOSPC408328-500MG
4-(4-Fluoro-benzenesulphonyl)-piperidine-1-carboxylic acid tert-butyl ester
4-(4-Fluoro-benzenesulphonyl)-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₆H₂₂FNO₄S
Molecular Weight: 343,414 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657077
CAS-Nummer: 226398-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR