3-(4-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-(((4-fluorophenyl)thio)methyl)piperidine-1-carboxylate

PC408331-500MG
APOSPC408331-500MGEA 0 EUR
APOSPC408331-500MG
3-(4-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
3-(4-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₄FNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD18837557
CAS-Nummer: 1289388-37-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR