3-(4-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC408332-500MG
APOSPC408332-500MGEA 430 EUR
APOSPC408332-500MG
3-(4-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
3-(4-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
Formel: C₁₂H₁₆FNS·ClH
Molecular Weight: 261,786 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837558
CAS-Nummer: 1289385-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR