3-(4-Fluoro-benzenesulphonylmethyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC408333-500MG
APOSPC408333-500MGEA 430 EUR
APOSPC408333-500MG
3-(4-Fluoro-benzenesulphonylmethyl)-piperidine hydrochloride
3-(4-Fluoro-benzenesulphonylmethyl)-piperidine hydrochloride
Formel: C₁₂H₁₆FNO₂S·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD18837559
CAS-Nummer: 1289388-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR