3-(4-Fluoro-benzenesulphinylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
PC408334-500MG
APOSPC408334-500MGEA
430
EUR
APOSPC408334-500MG
3-(4-Fluoro-benzenesulphinylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
3-(4-Fluoro-benzenesulphinylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Formel:
C₁₇H₂₄FNO₃S Molecular Weight: 341,441 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18837560 CAS-Nummer: 1289385-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...