(S)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408336-500MG
APOSPC408336-500MGEA 430 EUR
APOSPC408336-500MG
(S)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
(S)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Formel: C₁₅H₂₀FNO₄S
Molecular Weight: 329,387 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837564
CAS-Nummer: 1289585-25-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR