1-(2-Fluoro-benzyl)-azetidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC408337-500MG
APOSPC408337-500MGEA 430 EUR
APOSPC408337-500MG
1-(2-Fluoro-benzyl)-azetidine-3-carboxylic acid
1-(2-Fluoro-benzyl)-azetidine-3-carboxylic acid
Formel: C₁₁H₁₂FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18837580
CAS-Nummer: 1289387-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR