N-(3-Fluoro-benzyl)-cyclohexane-1,2-diamine hydrochloride
Lieferant: Apollo Scientific
PC408338-500MG
APOSPC408338-500MGEA
430
EUR
APOSPC408338-500MG
N-(3-Fluoro-benzyl)-cyclohexane-1,2-diamine hydrochloride
N-(3-Fluoro-benzyl)-cyclohexane-1,2-diamine hydrochloride
Formel:
C₁₃H₁₉FN₂·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD21098467 CAS-Nummer: 1353954-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...