N,N'-Bis-(3-fluoro-benzyl)-cyclohexane-1,4-diamine

Lieferant: Apollo Scientific

PC408339-500MG
APOSPC408339-500MGEA 430 EUR
APOSPC408339-500MG
N,N'-Bis-(3-fluoro-benzyl)-cyclohexane-1,4-diamine
N,N'-Bis-(3-fluoro-benzyl)-cyclohexane-1,4-diamine
Formel: C₂₀H₂₄F₂N₂
Molecular Weight: 330,415 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098468
CAS-Nummer: 1353987-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR