(1R,4R)-N-(2-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride

Lieferant: Apollo Scientific

PC408341-500MG
APOSPC408341-500MGEA 430 EUR
APOSPC408341-500MG
(1R,4R)-N-(2-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
(1R,4R)-N-(2-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
Formel: C₁₃H₁₉FN₂·ClH
Molecular Weight: 258,763 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098476
CAS-Nummer: 1366386-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR