4-(4-Fluoro-benzenesulphinylmethyl)-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408342-500MG
APOSPC408342-500MGEA 495 EUR
APOSPC408342-500MG
4-(4-Fluoro-benzenesulphinylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
4-(4-Fluoro-benzenesulphinylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₄FNO₃S
Molecular Weight: 341,441 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098479
CAS-Nummer: 1353989-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR