4-(4-Fluoro-benzenesulphonyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC408343-500MG
APOSPC408343-500MGEA 495 EUR
APOSPC408343-500MG
4-(4-Fluoro-benzenesulphonyl)-piperidine hydrochloride
4-(4-Fluoro-benzenesulphonyl)-piperidine hydrochloride
Formel: C₁₁H₁₄FNO₂S·ClH
Molecular Weight: 279,759 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837553
CAS-Nummer: 105283-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR