(R)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408344-500MG
APOSPC408344-500MGEA 495 EUR
APOSPC408344-500MG
(R)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
(R)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Formel: C₁₅H₂₀FNO₄S
Molecular Weight: 329,387 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098484
CAS-Nummer: 1354010-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR