2-(4-Fluoro-benzenesulphonylmethyl)-pyrrolidine hydrochloride

Lieferant: Apollo Scientific

PC408345-500MG
APOSPC408345-500MGEA 495 EUR
APOSPC408345-500MG
2-(4-Fluoro-benzenesulphonylmethyl)-pyrrolidine hydrochloride
2-(4-Fluoro-benzenesulphonylmethyl)-pyrrolidine hydrochloride
Formel: C₁₁H₁₄FNO₂S·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD21098491
CAS-Nummer: 1353977-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR