2-(4-Fluoro-benzenesulphonylmethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
PC408346-500MG
APOSPC408346-500MGEA
495
EUR
APOSPC408346-500MG
2-(4-Fluoro-benzenesulphonylmethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
2-(4-Fluoro-benzenesulphonylmethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Formel:
C₁₆H₂₂FNO₄S Molecular Weight: 343,414 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21098492 CAS-Nummer: 1353973-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...