2-(4-Fluoro-phenylsulphanylmethyl)-pyrrolidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 2-(((4-Fluorophenyl)thio)methyl)pyrrolidine hydrochloride

PC408347-500MG
APOSPC408347-500MGEA 430 EUR
APOSPC408347-500MG
2-(4-Fluoro-phenylsulphanylmethyl)-pyrrolidine hydrochloride
2-(4-Fluoro-phenylsulphanylmethyl)-pyrrolidine hydrochloride
Formel: C₁₁H₁₄FNS·ClH
Molecular Weight: 247,76 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098493
CAS-Nummer: 1353989-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR